Learn More
Meso Scale Discovery UPLEX HUM FGF basic ASY384well
SDP
Catalog No. 5000080517
Encompass
The Meso Scale Discovery U-PLEX Human FGF Basic 384-Well Assay is a multiplex immunoassay that measures FGF basic in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay can measure FGF basic levels between 0 06 - 2500 pg/mL and has high specificity with minimal cross-reactivity It requires just 25 L of sample per well and the 384-well plate allows high-throughput analysis The assay takes approximately 3 hours to complete
Catalog No. 50-000-80517 Supplier Meso Scale Discovery Supplier No. K251X7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FGF Basic 384-Well Assay is a multiplex immunoassay that measures FGF basic in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay can measure FGF basic levels between 0 06 - 2500 pg/mL and has high specificity with minimal cross-reactivity It requires just 25 L of sample per well and the 384-well plate allows high-throughput analysis The assay takes approximately 3 hours to complete