Learn More
Meso Scale Discovery UPLEX HUM FGF21 ASY384well
SDP
Catalog No. 5000080744
Encompass
The Meso Scale Discovery U-PLEX Human FGF-21 384-Well Assay Kit is a multiplex immunoassay that measures FGF-21 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows high-throughput analysis of FGF-21 levels with a small sample volume requirement of 25 L The kit has a typical assay range of 1 53 - 10 000 pg/mL and a lower limit of detection of 0 74 pg/mL for FGF-21
Catalog No. 50-000-80744 Supplier Meso Scale Discovery Supplier No. K2515WK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FGF-21 384-Well Assay Kit is a multiplex immunoassay that measures FGF-21 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows high-throughput analysis of FGF-21 levels with a small sample volume requirement of 25 L The kit has a typical assay range of 1 53 - 10 000 pg/mL and a lower limit of detection of 0 74 pg/mL for FGF-21