Learn More
Meso Scale Discovery UPLEX HUM FGF23 ASY384well
SDP
Catalog No. 5000078633
Encompass
The Meso Scale Discovery U-PLEX Human FGF-23 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring FGF-23 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Requiring only 25 L it s ideal for high throughput screening accurately determining FGF-23 levels across a wide range
Catalog No. 50-000-78633 Supplier Meso Scale Discovery Supplier No. K2516EK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FGF-23 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring FGF-23 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Requiring only 25 L it s ideal for high throughput screening accurately determining FGF-23 levels across a wide range