Learn More
Meso Scale Discovery UPLEX HUM FGF23 ASY384well
SDP
Catalog No. 5000081101
Encompass
The Meso Scale Discovery U-PLEX Human FGF-23 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human fibroblast growth factor 23 (FGF-23) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 384-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents It measures FGF-23 concentrations from 0 61 to 2500 pg/mL with high precision and accuracy
Catalog No. 50-000-81101 Supplier Meso Scale Discovery Supplier No. K2516EK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FGF-23 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human fibroblast growth factor 23 (FGF-23) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 384-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents It measures FGF-23 concentrations from 0 61 to 2500 pg/mL with high precision and accuracy