Learn More
Meso Scale Discovery UPLEX HUM Fractalkine ASY
SDP
Catalog No. 5000081433
Encompass
The Meso Scale Discovery U-PLEX Human Fractalkine Assay Kit is a multiplex immunoassay that measures fractalkine levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81433 Supplier Meso Scale Discovery Supplier No. K151VCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Fractalkine Assay Kit is a multiplex immunoassay that measures fractalkine levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL