Learn More
Meso Scale Discovery UPLEX HUM Fractalkine ASY384
SDP
Catalog No. 5000080719
Encompass
The Meso Scale Discovery U-PLEX Human Fractalkine 384-well Assay is a multiplex immunoassay kit for quantifying fractalkine in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 384-well plate analyzing up to 80 samples per plate With a sensitivity of 0 04 pg/mL it requires 50 L of sample per well The kit includes a plate with capture antibodies detection antibodies calibrators and controls offering a rapid sensitive and reproducible method for measuring fractalkine levels
Catalog No. 50-000-80719 Supplier Meso Scale Discovery Supplier No. K251VCK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Fractalkine 384-well Assay is a multiplex immunoassay kit for quantifying fractalkine in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 384-well plate analyzing up to 80 samples per plate With a sensitivity of 0 04 pg/mL it requires 50 L of sample per well The kit includes a plate with capture antibodies detection antibodies calibrators and controls offering a rapid sensitive and reproducible method for measuring fractalkine levels