Learn More
Meso Scale Discovery UPLEX HUM Fractalkine ASYQP
SDP
Catalog No. 5000079637
Encompass
The U-PLEX Human Fractalkine Assay (K151VCK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure fractalkine (CX3CL1) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying this important chemokine involved in immune cell recruitment and inflammation The kit includes pre-coated plates detection antibodies and calibrators for reliable reproducible results in research applications studying neuroinflammation cardiovascular disease and immune responses
Catalog No. 50-000-79637 Supplier Meso Scale Discovery Supplier No. K151VCK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human Fractalkine Assay (K151VCK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure fractalkine (CX3CL1) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying this important chemokine involved in immune cell recruitment and inflammation The kit includes pre-coated plates detection antibodies and calibrators for reliable reproducible results in research applications studying neuroinflammation cardiovascular disease and immune responses