Learn More
Meso Scale Discovery UPLEX HUM Galectin9 ASY384
SDP
Catalog No. 5000080987
Encompass
The Meso Scale Discovery Galectin-9 Kit is an electrochemiluminescent immunoassay for measuring Galectin-9 in serum plasma and cell culture supernatants It includes a 96-well plate with a capture antibody a detection antibody with an electrochemiluminescent label calibrators and buffers The assay s sensitivity is 0 3 ng/mL with a range of 0 3-1000 ng/mL It offers high sensitivity and a wide dynamic range taking about 3 5 hours to run The kit supports research on Galectin-9 s role in immune function and inflammation
Catalog No. 50-000-80987 Supplier Meso Scale Discovery Supplier No. K251AGWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Galectin-9 Kit is an electrochemiluminescent immunoassay for measuring Galectin-9 in serum plasma and cell culture supernatants It includes a 96-well plate with a capture antibody a detection antibody with an electrochemiluminescent label calibrators and buffers The assay s sensitivity is 0 3 ng/mL with a range of 0 3-1000 ng/mL It offers high sensitivity and a wide dynamic range taking about 3 5 hours to run The kit supports research on Galectin-9 s role in immune function and inflammation