Learn More
Meso Scale Discovery UPLEX HUM Galectin9 ASY384well
SDP
Catalog No. 5000080818
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of Galectin-9 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of Galectin-9 an important biomarker with a quick 3 5 hour protocol
Catalog No. 50-000-80818 Supplier Meso Scale Discovery Supplier No. K251AGWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Galectin-9 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of Galectin-9 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of Galectin-9 an important biomarker with a quick 3 5 hour protocol