Learn More
Meso Scale Discovery UPLEX HUM Galectin9 ASYQP
SDP
Catalog No. 5000081586
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of galectin-9 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals It allows rapid and accurate quantification of galectin-9 levels which can aid in understanding immune response regulation
Catalog No. 50-000-81586 Supplier Meso Scale Discovery Supplier No. K151AGWK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of galectin-9 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals It allows rapid and accurate quantification of galectin-9 levels which can aid in understanding immune response regulation