Learn More
Meso Scale Discovery UPLEX HUM GCSF ASY384well
SDP
Catalog No. 5000079472
Encompass
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media The assay uses MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure G-CSF over a range of 156 - 20 000 pg/mL with an LLOQ of 64 5 pg/mL
Catalog No. 50-000-79472 Supplier Meso Scale Discovery Supplier No. K251VGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media The assay uses MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure G-CSF over a range of 156 - 20 000 pg/mL with an LLOQ of 64 5 pg/mL