Learn More
Meso Scale Discovery UPLEX HUM Ghrelin active ASYQP
SDP
Catalog No. 5000080214
Encompass
Meso Scale Discovery offers ghrelin (active) assay kits for quantitative measurement of active ghrelin in biological samples These electrochemiluminescence-based immunoassays provide high sensitivity detection of the 28-amino acid peptide hormone that regulates appetite and energy balance The kits are designed for research applications in metabolism obesity and endocrine studies featuring multiplex capabilities and requiring minimal sample volumes for accurate quantification in serum plasma and other biological matrices
Catalog No. 50-000-80214 Supplier Meso Scale Discovery Supplier No. K1516KK22
Meso Scale Discovery offers ghrelin (active) assay kits for quantitative measurement of active ghrelin in biological samples These electrochemiluminescence-based immunoassays provide high sensitivity detection of the 28-amino acid peptide hormone that regulates appetite and energy balance The kits are designed for research applications in metabolism obesity and endocrine studies featuring multiplex capabilities and requiring minimal sample volumes for accurate quantification in serum plasma and other biological matrices