Learn More
Meso Scale Discovery UPLEX HUM Ghrelin TTL ASY384
SDP
Supplier:  Meso Scale Discovery K2515XK2
Encompass
The Meso Scale Discovery U-PLEX Human Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay that measures total ghrelin in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a sensitivity of 0 3 pg/mL and a 7-point standard curve from 0 3-2000 pg/mL It requires a 2 hour incubation time and 10 L sample size This assay kit offers a reproducible and robust immunoassay for measuring total ghrelin
Catalog No. 50-000-78623
Add to Cart
Request a Quote