Learn More
Meso Scale Discovery UPLEX HUM Ghrelin TTL ASY384
SDP
Catalog No. 5000081080
Encompass
The Meso Scale Discovery U-PLEX Human Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay that measures total ghrelin levels in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a sensitivity of 0 3 pg/mL and a 7-point standard curve from 0 3-2000 pg/mL Each kit provides reagents for 2 x 384-well plates when used as directed This assay kit offers a rapid reproducible method for quantifying total ghrelin levels in various human sample types
Catalog No. 50-000-81080 Supplier Meso Scale Discovery Supplier No. K2515XK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay that measures total ghrelin levels in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a sensitivity of 0 3 pg/mL and a 7-point standard curve from 0 3-2000 pg/mL Each kit provides reagents for 2 x 384-well plates when used as directed This assay kit offers a rapid reproducible method for quantifying total ghrelin levels in various human sample types