Learn More
Meso Scale Discovery UPLEX HUM GIP active ASYQP
SDP
Catalog No. 5000082443
Encompass
The U-PLEX Human GIP Active Assay (K1516NK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure active glucose-dependent insulinotropic polypeptide (GIP) levels in biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for detecting the bioactive form of GIP an incretin hormone involved in glucose homeostasis and insulin regulation The kit utilizes MSD s proprietary U-PLEX platform technology for reliable quantitative analysis in research applications studying metabolic disorders and diabetes
Catalog No. 50-000-82443 Supplier Meso Scale Discovery Supplier No. K1516NK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human GIP Active Assay (K1516NK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure active glucose-dependent insulinotropic polypeptide (GIP) levels in biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for detecting the bioactive form of GIP an incretin hormone involved in glucose homeostasis and insulin regulation The kit utilizes MSD s proprietary U-PLEX platform technology for reliable quantitative analysis in research applications studying metabolic disorders and diabetes