Learn More
Meso Scale Discovery UPLEX HUM GIP TTL ASY384well
SDP
Catalog No. 5000080903
Encompass
The Meso Scale Discovery U-PLEX Human GIP Total 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total GIP in human serum plasma (EDTA heparin) and cell culture supernatants The assay has a detection range of 0 06 - 1000 pg/mL and includes a plate coated with capture antibody detection antibody labeled with an electrochemiluminescent compound and calibrator The kit provides components for one 384-well plate allowing measurement of up to 352 samples in duplicate
Catalog No. 50-000-80903 Supplier Meso Scale Discovery Supplier No. K2515SK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GIP Total 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total GIP in human serum plasma (EDTA heparin) and cell culture supernatants The assay has a detection range of 0 06 - 1000 pg/mL and includes a plate coated with capture antibody detection antibody labeled with an electrochemiluminescent compound and calibrator The kit provides components for one 384-well plate allowing measurement of up to 352 samples in duplicate