Learn More
Meso Scale Discovery UPLEX HUM GITR/TNFRSF18 ASY
SDP
Catalog No. 5000078785
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human GITR in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and compatibility with small sample volumes The assay can be multiplexed with other U-PLEX biomarker assays for efficient and customizable biomarker profiling With a rapid 3 5 hour protocol and minimal hands-on time this assay allows for high-throughput quantification of GITR levels
Catalog No. 50-000-78785 Supplier Meso Scale Discovery Supplier No. K151Y7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human GITR in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and compatibility with small sample volumes The assay can be multiplexed with other U-PLEX biomarker assays for efficient and customizable biomarker profiling With a rapid 3 5 hour protocol and minimal hands-on time this assay allows for high-throughput quantification of GITR levels