Learn More
Meso Scale Discovery UPLEX HUM GITR/TNFRSF18 ASYQP
SDP
Catalog No. 5000078932
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume It quantifies human GITR over a wide range and is validated for research use to study GITR biology The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals for one 96-well plate
Catalog No. 50-000-78932 Supplier Meso Scale Discovery Supplier No. K151Y7K21
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume It quantifies human GITR over a wide range and is validated for research use to study GITR biology The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals for one 96-well plate