Learn More
Meso Scale Discovery UPLEX HUM GITR/TNFRSF18 ASYQP
SDP
Catalog No. 5000080110
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay panel that measures GITR in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of GITR which plays an important role in modulating T cell activation and immune responses With minimal hands-on time and rapid results this assay enables efficient profiling of GITR expression
Catalog No. 50-000-80110 Supplier Meso Scale Discovery Supplier No. K151Y7K22
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay panel that measures GITR in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of GITR which plays an important role in modulating T cell activation and immune responses With minimal hands-on time and rapid results this assay enables efficient profiling of GITR expression