Learn More
Meso Scale Discovery UPLEX HUM GITRL/TNFSF18 ASY384
SDP
Catalog No. 5000080688
Encompass
The GITRL/TNFSF18 Human ELISA Kit from Meso Scale Discovery is an enzyme-linked immunosorbent assay for the quantitative measurement of GITRL/TNFSF18 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GITRL/TNFSF18 capture antibody a detection antibody and an MSD SULFO-TAG label for electrochemiluminescent detection The assay has a sensitivity of 0 3 pg/mL and a range of 0 3-1500 pg/mL The total assay time is 3 5 hours This product can be used to measure GITRL/TNFSF18 levels in research related to autoimmune diseases inflammation and immune system regulation
Catalog No. 50-000-80688 Supplier Meso Scale Discovery Supplier No. K251S7K4
Add to Cart
Add to Cart
Request a Quote
The GITRL/TNFSF18 Human ELISA Kit from Meso Scale Discovery is an enzyme-linked immunosorbent assay for the quantitative measurement of GITRL/TNFSF18 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GITRL/TNFSF18 capture antibody a detection antibody and an MSD SULFO-TAG label for electrochemiluminescent detection The assay has a sensitivity of 0 3 pg/mL and a range of 0 3-1500 pg/mL The total assay time is 3 5 hours This product can be used to measure GITRL/TNFSF18 levels in research related to autoimmune diseases inflammation and immune system regulation