Learn More
Meso Scale Discovery UPLEX HUM GITRL/TNFSF18 ASY384
SDP
Catalog No. 5000080566
Encompass
The Meso Scale Discovery GITRL (TNFSF18) Human Proinflammatory 2 Kit is a highly sensitive immunoassay for the quantitative measurement of GITRL in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GITRL capture antibody a ruthenylated GITRL detection antibody assay diluent calibrator diluent wash buffer read buffer and a human GITRL calibrator The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This product can accurately quantify GITRL levels from 0 156 pg/mL to 10 000 pg/mL
Catalog No. 50-000-80566 Supplier Meso Scale Discovery Supplier No. K251S7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery GITRL (TNFSF18) Human Proinflammatory 2 Kit is a highly sensitive immunoassay for the quantitative measurement of GITRL in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GITRL capture antibody a ruthenylated GITRL detection antibody assay diluent calibrator diluent wash buffer read buffer and a human GITRL calibrator The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This product can accurately quantify GITRL levels from 0 156 pg/mL to 10 000 pg/mL