Learn More
Meso Scale Discovery UPLEX HUM GITRL/TNFSF18 ASYQP
SDP
Catalog No. 5000081686
Encompass
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITRL in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive measurements with minimal sample volume The assay is automatable on the QuickPlex SQ 120 and detects as low as 0 04 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents
Catalog No. 50-000-81686 Supplier Meso Scale Discovery Supplier No. K151S7K22
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITRL in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive measurements with minimal sample volume The assay is automatable on the QuickPlex SQ 120 and detects as low as 0 04 ng/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents