Learn More
Meso Scale Discovery UPLEX HUM GLP1 active ASYQP
SDP
Catalog No. 5000081711
Encompass
MSD s GLP-1 Active assay is a multiplex immunoassay platform designed for quantitative measurement of active GLP-1 hormone levels in biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for research applications studying glucose metabolism diabetes and incretin hormone pathways The platform enables efficient sample processing with reduced sample volumes and provides reliable reproducible results for pharmaceutical and academic research settings
Catalog No. 50-000-81711 Supplier Meso Scale Discovery Supplier No. K1516LK21
MSD s GLP-1 Active assay is a multiplex immunoassay platform designed for quantitative measurement of active GLP-1 hormone levels in biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for research applications studying glucose metabolism diabetes and incretin hormone pathways The platform enables efficient sample processing with reduced sample volumes and provides reliable reproducible results for pharmaceutical and academic research settings