Learn More
Meso Scale Discovery UPLEX HUM GLP1 TTL ASY384well
SDP
Catalog No. 5000078631
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 Total 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in human serum plasma and cell culture supernatants The assay uses MSD GOLD technology and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78631 Supplier Meso Scale Discovery Supplier No. K2515UK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GLP-1 Total 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in human serum plasma and cell culture supernatants The assay uses MSD GOLD technology and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL