Learn More
Meso Scale Discovery UPLEX HUM GLP1 TTL ASY384well
SDP
Catalog No. 5000080778
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 Total 384-Well Assay is an electrochemiluminescent immunoassay designed to measure total GLP-1 levels in human serum plasma and other biological samples This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a broad dynamic range of up to 4 logs and requires just 12 5 L of sample per well It is suitable for high-throughput screening and provides quantitative data on GLP-1 levels that can aid in research on diabetes obesity and metabolic disorders
Catalog No. 50-000-80778 Supplier Meso Scale Discovery Supplier No. K2515UK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GLP-1 Total 384-Well Assay is an electrochemiluminescent immunoassay designed to measure total GLP-1 levels in human serum plasma and other biological samples This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a broad dynamic range of up to 4 logs and requires just 12 5 L of sample per well It is suitable for high-throughput screening and provides quantitative data on GLP-1 levels that can aid in research on diabetes obesity and metabolic disorders