Learn More
Meso Scale Discovery UPLEX HUM GLP1 TTL ASYQP
SDP
Catalog No. 5000078411
Encompass
The U-PLEX Human GLP-1 Total Assay (K1515UK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure total GLP-1 levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying glucagon-like peptide-1 a key incretin hormone involved in glucose homeostasis and diabetes research The kit includes pre-coated plates detection antibodies and calibrators for reliable reproducible results in clinical and research applications
Catalog No. 50-000-78411 Supplier Meso Scale Discovery Supplier No. K1515UK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human GLP-1 Total Assay (K1515UK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure total GLP-1 levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying glucagon-like peptide-1 a key incretin hormone involved in glucose homeostasis and diabetes research The kit includes pre-coated plates detection antibodies and calibrators for reliable reproducible results in clinical and research applications