Learn More
Meso Scale Discovery UPLEX HUM GLPinactive ASY384
SDP
Catalog No. 5000080909
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 (Inactive) 384-Well Assay Kit is designed to measure GLP-1 (inactive) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time to results This multiplex assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls It enables quantification of GLP-1 (inactive) levels with a testing time of just 3 5 hours With a lower limit of detection of 49 pg/mL this assay kit offers superb analytical performance for measuring GLP-1 (inactive)
Catalog No. 50-000-80909 Supplier Meso Scale Discovery Supplier No. K2515VK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GLP-1 (Inactive) 384-Well Assay Kit is designed to measure GLP-1 (inactive) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time to results This multiplex assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls It enables quantification of GLP-1 (inactive) levels with a testing time of just 3 5 hours With a lower limit of detection of 49 pg/mL this assay kit offers superb analytical performance for measuring GLP-1 (inactive)