Learn More
Meso Scale Discovery UPLEX HUM GLPinactive ASY384
SDP
Catalog No. 5000081100
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 (Inactive) 384-Well Assay Kit is a multiplex immunoassay that measures GLP-1 (inactive) in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient analysis of large sample sets The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and substrate It can detect GLP-1 (inactive) concentrations as low as 17 2 pg/mL with an upper limit of 50 000 pg/mL
Catalog No. 50-000-81100 Supplier Meso Scale Discovery Supplier No. K2515VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GLP-1 (Inactive) 384-Well Assay Kit is a multiplex immunoassay that measures GLP-1 (inactive) in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient analysis of large sample sets The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and substrate It can detect GLP-1 (inactive) concentrations as low as 17 2 pg/mL with an upper limit of 50 000 pg/mL