Learn More
Meso Scale Discovery UPLEX HUM Glucagon ASY384well
SDP
Catalog No. 5000080905
Encompass
The Meso Scale Discovery U-PLEX Human Glucagon 384-Well Assay Kit is a multiplex immunoassay that measures glucagon levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of glucagon The assay has a detection range of 1 53 - 1000 pg/mL and includes a standard curve for quantitative analysis The total assay time is approximately 3 5 hours This assay kit can be used for research applications to study glucagon biology and diabetes
Catalog No. 50-000-80905 Supplier Meso Scale Discovery Supplier No. K2515YK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Glucagon 384-Well Assay Kit is a multiplex immunoassay that measures glucagon levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of glucagon The assay has a detection range of 1 53 - 1000 pg/mL and includes a standard curve for quantitative analysis The total assay time is approximately 3 5 hours This assay kit can be used for research applications to study glucagon biology and diabetes