Learn More
Meso Scale Discovery UPLEX HUM gp130 soluble ASY
SDP
Catalog No. 5000082270
Encompass
The Meso Scale Discovery U-PLEX Human GP130 Soluble Assay is an electrochemiluminescent immunoassay for the quantitative measurement of soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity for soluble GP130 with no detectable cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrators and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-82270 Supplier Meso Scale Discovery Supplier No. K151P7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GP130 Soluble Assay is an electrochemiluminescent immunoassay for the quantitative measurement of soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity for soluble GP130 with no detectable cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrators and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL