Learn More
Meso Scale Discovery UPLEX HUM gp130 soluble ASY384
SDP
Catalog No. 5000080464
Encompass
The Meso Scale Discovery U-PLEX Human GP130 Soluble 384-Well Assay is a 384-well electrochemiluminescent immunoassay designed to measure soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of soluble GP130 with a broad dynamic range This assay requires a low sample volume of 25 L and has a total assay time of approximately 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent
Catalog No. 50-000-80464 Supplier Meso Scale Discovery Supplier No. K251P7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GP130 Soluble 384-Well Assay is a 384-well electrochemiluminescent immunoassay designed to measure soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of soluble GP130 with a broad dynamic range This assay requires a low sample volume of 25 L and has a total assay time of approximately 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent