Learn More
Meso Scale Discovery UPLEX HUM gp130 soluble ASYQP
SDP
Catalog No. 5000079267
Encompass
The Meso Scale Discovery U-PLEX Human GP130 Soluble Assay Kit is a multiplex immunoassay that measures soluble GP130 in human serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can test up to 80 samples in duplicate and has a lower limit of detection of 0 04 ng/mL
Catalog No. 50-000-79267 Supplier Meso Scale Discovery Supplier No. K151P7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GP130 Soluble Assay Kit is a multiplex immunoassay that measures soluble GP130 in human serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can test up to 80 samples in duplicate and has a lower limit of detection of 0 04 ng/mL