Learn More
Meso Scale Discovery UPLEX HUM gp130 soluble ASYQP
SDP
Catalog No. 5000079607
Encompass
The Meso Scale Discovery U-PLEX Human GP130 (Soluble) Assay Kit is designed for the quantitative measurement of soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for soluble GP130 with no cross-reactivity to other soluble receptors The kit includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrators and controls to generate a standard curve Sample incubation time is just 3 5 hours The kit can test up to 80 samples in duplicate
Catalog No. 50-000-79607 Supplier Meso Scale Discovery Supplier No. K151P7K22
The Meso Scale Discovery U-PLEX Human GP130 (Soluble) Assay Kit is designed for the quantitative measurement of soluble GP130 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for soluble GP130 with no cross-reactivity to other soluble receptors The kit includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrators and controls to generate a standard curve Sample incubation time is just 3 5 hours The kit can test up to 80 samples in duplicate