Learn More
Meso Scale Discovery UPLEX HUM Granzyme A ASY384
SDP
Catalog No. 5000079404
Encompass
The Meso Scale Discovery U-PLEX Human Granzyme A 384-Well Assay Kit is a multiplex immunoassay that measures Granzyme A levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of Granzyme A The assay has high sensitivity wide dynamic range and requires low sample volume This assay kit allows quantification of the cytotoxic T cell and NK cell mediator Granzyme A involved in targeted cell apoptosis It can be used for monitoring immune system function and immunotherapy response
Catalog No. 50-000-79404 Supplier Meso Scale Discovery Supplier No. K251G8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Granzyme A 384-Well Assay Kit is a multiplex immunoassay that measures Granzyme A levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of Granzyme A The assay has high sensitivity wide dynamic range and requires low sample volume This assay kit allows quantification of the cytotoxic T cell and NK cell mediator Granzyme A involved in targeted cell apoptosis It can be used for monitoring immune system function and immunotherapy response