Learn More
Meso Scale Discovery UPLEX HUM Granzyme A ASY384
SDP
Catalog No. 5000079479
Encompass
The Meso Scale Discovery U-PLEX Human Granzyme A 384-Well Assay Kit is designed for the quantitative measurement of Granzyme A in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for 384 tests The assay has a typical sensitivity of 0 04 pg/mL and an assay range from 0 08-500 pg/mL This multiplex immunoassay allows high-throughput profiling of Granzyme A with low sample volume requirements
Catalog No. 50-000-79479 Supplier Meso Scale Discovery Supplier No. K251G8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Granzyme A 384-Well Assay Kit is designed for the quantitative measurement of Granzyme A in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for 384 tests The assay has a typical sensitivity of 0 04 pg/mL and an assay range from 0 08-500 pg/mL This multiplex immunoassay allows high-throughput profiling of Granzyme A with low sample volume requirements