Learn More
Meso Scale Discovery UPLEX HUM GRO ASY384well
SDP
Catalog No. 5000080659
Encompass
The Meso Scale Discovery U-PLEX Human GRO-alpha 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of GRO-alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluents optimized for use with MSD readers It allows high-throughput analysis of GRO-alpha levels with minimal hands-on time
Catalog No. 50-000-80659 Supplier Meso Scale Discovery Supplier No. K251UXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human GRO-alpha 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of GRO-alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluents optimized for use with MSD readers It allows high-throughput analysis of GRO-alpha levels with minimal hands-on time