Learn More
Meso Scale Discovery UPLEX HUM HAVCR2/TIM3 ASY384
SDP
Catalog No. 5000078529
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human TIM-3 and other biomarkers in serum plasma or cell culture supernatants The electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a typical sensitivity of 0 3 pg/mL and requires just 12 5 L of sample per well The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators to generate a standard curve
Catalog No. 50-000-78529 Supplier Meso Scale Discovery Supplier No. K251E8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human TIM-3 and other biomarkers in serum plasma or cell culture supernatants The electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a typical sensitivity of 0 3 pg/mL and requires just 12 5 L of sample per well The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators to generate a standard curve