Learn More
Meso Scale Discovery UPLEX HUM HAVCR2/TIM3 ASYQP
SDP
Catalog No. 5000079694
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human TIM-3 and other biomarkers This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from as little as 25 L of sample With minimal sample consumption rapid turnaround time and standard curves spanning wide dynamic ranges this assay kit offers an efficient way to measure TIM-3 and other immune checkpoint targets from serum plasma cell culture supernatants and other biological matrices
Catalog No. 50-000-79694 Supplier Meso Scale Discovery Supplier No. K151E8K22
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human TIM-3 and other biomarkers This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from as little as 25 L of sample With minimal sample consumption rapid turnaround time and standard curves spanning wide dynamic ranges this assay kit offers an efficient way to measure TIM-3 and other immune checkpoint targets from serum plasma cell culture supernatants and other biological matrices