Learn More
Meso Scale Discovery UPLEX HUM HAVCR2/TIM3 ASYQP
SDP
Catalog No. 5000082095
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit for detecting and quantifying human HAVCR2 (TIM-3) in various biological samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay supports efficient reproducible and sensitive measurement of HAVCR2 (TIM-3) levels crucial for immunology and oncology research
Catalog No. 50-000-82095 Supplier Meso Scale Discovery Supplier No. K151E8K21
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit for detecting and quantifying human HAVCR2 (TIM-3) in various biological samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay supports efficient reproducible and sensitive measurement of HAVCR2 (TIM-3) levels crucial for immunology and oncology research