Learn More
Meso Scale Discovery UPLEX HUM HAVCR2/TIMASY384
SDP
Catalog No. 5000080751
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) 384-well Assay Kit is a multiplex immunoassay that measures human TIM-3 in cell culture supernatants serum plasma and other biological samples This kit uses electrochemiluminescence (ECL) detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity wide dynamic range and requires low sample volume This product can help researchers study immune checkpoint pathways and mechanisms and aid in the discovery of therapeutics targeting TIM-3
Catalog No. 50-000-80751 Supplier Meso Scale Discovery Supplier No. K251E8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) 384-well Assay Kit is a multiplex immunoassay that measures human TIM-3 in cell culture supernatants serum plasma and other biological samples This kit uses electrochemiluminescence (ECL) detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity wide dynamic range and requires low sample volume This product can help researchers study immune checkpoint pathways and mechanisms and aid in the discovery of therapeutics targeting TIM-3