Learn More
Meso Scale Discovery UPLEX HUM HVEM/TNFRSF14 ASY
SDP
Catalog No. 5000081649
Encompass
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human HVEM in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for human HVEM with a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL The assay requires 50 L of sample per well and can be read on MSD instrument platforms This product allows for high-throughput multiplexed immunoassays to advance research of the HVEM pathway
Catalog No. 50-000-81649 Supplier Meso Scale Discovery Supplier No. K151AGXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human HVEM in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for human HVEM with a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL The assay requires 50 L of sample per well and can be read on MSD instrument platforms This product allows for high-throughput multiplexed immunoassays to advance research of the HVEM pathway