Learn More
Meso Scale Discovery UPLEX HUM HVEM/TNFRSF14 ASY384
SDP
Catalog No. 5000080235
Encompass
The Human Vascular Endothelial Cell Adhesion Molecule (HVEM/TNFRSF14) ELISA Kit is an in vitro assay for measuring human HVEM/TNFRSF14 in serum plasma and other fluids It uses an antibody specific for HVEM/TNFRSF14 on a 96-well plate Samples bind to the immobilized antibody followed by a biotinylated anti-HVEM/TNFRSF14 antibody After washing HRP-conjugated streptavidin is added A TMB substrate develops color proportional to HVEM/TNFRSF14 and a stop solution changes the color for measurement at 450 nm
Catalog No. 50-000-80235 Supplier Meso Scale Discovery Supplier No. K251AGXK2
Add to Cart
Add to Cart
Request a Quote
The Human Vascular Endothelial Cell Adhesion Molecule (HVEM/TNFRSF14) ELISA Kit is an in vitro assay for measuring human HVEM/TNFRSF14 in serum plasma and other fluids It uses an antibody specific for HVEM/TNFRSF14 on a 96-well plate Samples bind to the immobilized antibody followed by a biotinylated anti-HVEM/TNFRSF14 antibody After washing HRP-conjugated streptavidin is added A TMB substrate develops color proportional to HVEM/TNFRSF14 and a stop solution changes the color for measurement at 450 nm