Learn More
Meso Scale Discovery UPLEX HUM HVEM/TNFRSF14 ASYQP
SDP
Catalog No. 5000081905
Encompass
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human Herpesvirus Entry Mediator (HVEM) in cell and tissue lysates using electrochemiluminescence (ECL) detection technology The assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve The assay measures HVEM an important immunomodulatory receptor belonging to the tumor necrosis factor receptor superfamily
Catalog No. 50-000-81905 Supplier Meso Scale Discovery Supplier No. K151AGXK22
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human Herpesvirus Entry Mediator (HVEM) in cell and tissue lysates using electrochemiluminescence (ECL) detection technology The assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve The assay measures HVEM an important immunomodulatory receptor belonging to the tumor necrosis factor receptor superfamily