Learn More
Meso Scale Discovery UPLEX HUM I309 ASYQP 25 PL
SDP
Catalog No. 5000080373
Encompass
The U-PLEX Human I-309 Assay (K151UYK) is a multiplex immunoassay platform by Meso Scale Discovery for quantitative measurement of I-309 (CCL1) chemokine levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers Designed for research applications in immunology and inflammation studies it provides reliable reproducible results for cytokine analysis in serum plasma and cell culture supernatants
Catalog No. 50-000-80373 Supplier Meso Scale Discovery Supplier No. K151UYK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human I-309 Assay (K151UYK) is a multiplex immunoassay platform by Meso Scale Discovery for quantitative measurement of I-309 (CCL1) chemokine levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers Designed for research applications in immunology and inflammation studies it provides reliable reproducible results for cytokine analysis in serum plasma and cell culture supernatants