Learn More
Meso Scale Discovery UPLEX HUM ICAM1 ASY 25 PL
SDP
Catalog No. 5000079655
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human ICAM-1 in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of ICAM-1 with high sensitivity and a broad dynamic range This product can help researchers study ICAM-1 levels in inflammation immune response and cardiovascular disease research The kit includes a 96-well plate calibrator diluent wash buffer read buffer 10X sample diluent and reagents for the assay and detection
Catalog No. 50-000-79655 Supplier Meso Scale Discovery Supplier No. K151E9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human ICAM-1 in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of ICAM-1 with high sensitivity and a broad dynamic range This product can help researchers study ICAM-1 levels in inflammation immune response and cardiovascular disease research The kit includes a 96-well plate calibrator diluent wash buffer read buffer 10X sample diluent and reagents for the assay and detection