Learn More
Meso Scale Discovery UPLEX HUM ICAM1 ASY384well
SDP
Catalog No. 5000081123
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 384-Well Assay Kit is a multiplex immunoassay that measures Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption It is available in a 384-well plate format includes reagents for up to 80 tests and has an assay time of 3 5 hours This assay kit can be used for biomarker research and drug discovery applications related to inflammation immune response and endothelial cell biology
Catalog No. 50-000-81123 Supplier Meso Scale Discovery Supplier No. K251E9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ICAM-1 384-Well Assay Kit is a multiplex immunoassay that measures Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption It is available in a 384-well plate format includes reagents for up to 80 tests and has an assay time of 3 5 hours This assay kit can be used for biomarker research and drug discovery applications related to inflammation immune response and endothelial cell biology