Learn More
Meso Scale Discovery UPLEX HUM ICAM1 ASYQP 25 PL
SDP
Catalog No. 5000082032
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human ICAM-1 in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2000 pg/mL This product can aid in understanding ICAM-1 levels related to inflammatory response
Catalog No. 50-000-82032 Supplier Meso Scale Discovery Supplier No. K151E9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human ICAM-1 in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2000 pg/mL This product can aid in understanding ICAM-1 levels related to inflammatory response