Learn More
Meso Scale Discovery UPLEX HUM ICOS ASY384well
SDP
Catalog No. 5000078377
Encompass
The Meso Scale Discovery U-PLEX Human ICOS Assay 384-Well Kit is a multiplex immunoassay that measures human ICOS in cell culture supernatants serum plasma and other biological samples It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of ICOS levels The assay requires 50 L of sample per well and can detect ICOS with a lower limit of detection of 49 pg/mL The kit includes a 384-well MULTI-ARRAY plate detection antibodies calibrators diluent read buffer wash buffer and plate seals This assay allows sensitive accurate and rapid analysis of ICOS
Catalog No. 50-000-78377 Supplier Meso Scale Discovery Supplier No. K251AHBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ICOS Assay 384-Well Kit is a multiplex immunoassay that measures human ICOS in cell culture supernatants serum plasma and other biological samples It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of ICOS levels The assay requires 50 L of sample per well and can detect ICOS with a lower limit of detection of 49 pg/mL The kit includes a 384-well MULTI-ARRAY plate detection antibodies calibrators diluent read buffer wash buffer and plate seals This assay allows sensitive accurate and rapid analysis of ICOS