Learn More
Meso Scale Discovery UPLEX HUM ICOSL/B7H2 ASYQP
SDP
Catalog No. 5000079522
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for efficient and reproducible analysis of ICOS and B7-H2 which play important roles in immune regulation
Catalog No. 50-000-79522 Supplier Meso Scale Discovery Supplier No. K151AHJK22
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for efficient and reproducible analysis of ICOS and B7-H2 which play important roles in immune regulation