Learn More
Meso Scale Discovery UPLEX HUM IL1 ASY 25 PL
SDP
Catalog No. 5000082025
Encompass
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay for measuring IL-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample volume needs The assay quantifies IL-1alpha from 0 06 to 2500 pg/mL with a 2-hour incubation It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator and control solutions optimized for reliable reproducible results
Catalog No. 50-000-82025 Supplier Meso Scale Discovery Supplier No. K151UNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay for measuring IL-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample volume needs The assay quantifies IL-1alpha from 0 06 to 2500 pg/mL with a 2-hour incubation It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator and control solutions optimized for reliable reproducible results